missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CTNNBL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555725
This item is not returnable.
View return policy
Description
Immunogen sequence: GVHNTLAIVE NMAEFRPEMC TEGAQQGLLQ WLLKRLKAKM PFDANKLYCS EVLAILLQDN DENRELLGEL DGIDVLLQQL SVFKRHNPST A Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
The protein encoded by this gene is a component of the pre-mRNA-processing factor 19-cell division cycle 5-like (PRP19-CDC5L) protein complex, which activates pre-mRNA splicing and is an integral part of the spliceosome. The encoded protein is also a nuclear localization sequence binding protein, and binds to activation-induced deaminase and is important for antibody diversification. This gene may also be associated with the development of obesity. Alternative splicing results in multiple transcript variants. A pseudogene of this gene has been defined on the X chromosome. Diseases associated with CTNNBL1 include Immunodeficiency 99 With Hypogammaglobulinemia And Autoimmune Cytopenias.
Specifications
| CTNNBL1 | |
| Polyclonal | |
| Unconjugated | |
| CTNNBL1 | |
| 5730471K09Rik; beta catenin-like 1; beta-catenin-like protein 1; C20orf33; catenin beta like 1; catenin, beta like 1; CTNNBL1; dJ633O20.1; NAP; nuclear associated protein; nuclear protein NAP; nuclear-associated protein; NYD-SP19; P14L; PP8304; RP5-1118M15.1; Testis development protein NYD-SP19 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56259 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WYA6 | |
| CTNNBL1 | |
| Recombinant protein corresponding to Human CTNNBL1. Recombinant protein control fragment (Product #RP-95289). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction