missing translation for 'onlineSavingsMsg'
Learn More

CTNNBL1 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18410596 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18410596 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18410596 Supplier Novus Biologicals Supplier No. NBP176543MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CTNNBL1 Polyclonal antibody specifically detects CTNNBL1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTNNBL1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias beta-catenin-like protein 1, C20orf33, catenin, beta like 1, dJ633O20.1, FLJ21108, NAPchromosome 20 open reading frame 33, nuclear associated protein, Nuclear-associated protein, NYD-SP19, P14, P14L, PP8304, Testis development protein NYD-SP19
Host Species Rabbit
Immunogen Antibody was raised against a 20 amino acid synthetic peptide near the carboxy terminus of human CTTNBL1. The immunogen is located within the last 50 amino acids of CTTNBL1. Amino Acid Squence: YAENIGDGRSPEFRENEQKR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 56259
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.