missing translation for 'onlineSavingsMsg'
Learn More

CTNNBL1 Antibody (5F1), Novus Biologicals™

Product Code. 18375669 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18375669 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18375669 Supplier Novus Biologicals Supplier No. H00056259M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CTNNBL1 Monoclonal antibody specifically detects CTNNBL1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTNNBL1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Sandwich ELISA
Classification Monoclonal
Clone 5F1
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Sandwich ELISA
Formulation 1x PBS, pH 7.4
Gene Accession No. BC036739
Gene Alias beta-catenin-like protein 1, C20orf33, catenin, beta like 1, dJ633O20.1, FLJ21108, NAPchromosome 20 open reading frame 33, nuclear associated protein, Nuclear-associated protein, NYD-SP19, P14, P14L, PP8304, Testis development protein NYD-SP19
Host Species Mouse
Immunogen CTNNBL1 (AAH36739, 210 a.a. ~ 311 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. EKHDMVRRGEIIDNDTEEEFYLRRLDAGLFVLQHICYIMAEICNANVPQIRQRVHQILNMRGSSIKIVRHIIKEYAENIGDGRSPEFRENEQKRILGLLENF
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 56259
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.