missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CTNNAL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522216
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, HeLa, HepG2. Predicted reactivity: Mouse (89%), Rat (89%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May modulate the Rho pathway signaling by providing a scaffold for the Lbc Rho guanine nucleotide exchange factor (ARHGEF1).
Specifications
| CTNNAL1 | |
| Polyclonal | |
| Unconjugated | |
| CTNNAL1 | |
| ACRP; AI616177; alpha2-catulin; alpha-catenin related protein; alpha-catenin-related protein; alpha-CATU; Alpha-catulin; AW545119; C86009; catenin (cadherin associated protein), alpha-like 1; catenin (cadherin-associated protein), alpha-like 1; catenin alpha like 1; catenin alpha-like 1; catenin alpha-like protein 1; Catnal1; CLLP; CTNNAL1; RP11-115J22.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54366, 8727 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| O88327, Q9UBT7 | |
| CTNNAL1 | |
| Recombinant protein encompassing a sequence within the center region of human CTNNAL1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction