missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTNNA3 Antibody (SR1012), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22396-100ul
This item is not returnable.
View return policy
Description
CTNNA3 Monoclonal antibody specifically detects CTNNA3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation
Specifications
| CTNNA3 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 29119 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| SR1012 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 | |
| Alpha T-catenin, alpha-catenin-like protein, alpha-T-catenin, Cadherin-associated protein, catenin (cadherin-associated protein), alpha 3, catenin alpha-3, MGC26194, MGC75041, VR22 | |
| A synthesized peptide derived from human CTNNA3 (Uniprot #: Q9UI47) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction