Learn More
Invitrogen™ Mouse CTLA-4 ELISA Kit
Description
The Mouse CTLA-4 ELISA quantitates Ms CTLA-4 in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms CTLA-4. Principle of the method The Mouse CTLA-4 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Specifications
Specifications
| Accession Number | P09793 |
| Assay Range | 4.92 to 1200 pg/mL |
| Assay Sensitivity | 5 pg/mL |
| Calibration Range | 5 pg/mL to 1200 pg/mL |
| Description | CTLA-4 ELISA Kit, 96 Assays, 1 Kit, Mouse Species, Chemokines and Receptors Protein Family, Immunology Research Discipline, Enzyme-Linked Immunosorbent Assay Designed for the Quantitation and Detection of Ctla-4 in Samples, Using a Microplate Reader |
| Detectable Analytes | CTLA-4 |
| Assay | Solid-phase sandwich ELISA |
| Conjugate | HRP |
| Detection Method | Colorimetric |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Show More |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.