missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CTLA-4 Antibody (Antitope patent anti-CTLA4) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
£162.00 - £695.00
Spécification
| Antigen | CTLA-4 |
|---|---|
| Clone | Antitope patent anti-CTLA4 |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|
30048738
|
Novus Biologicals™
NBP3-28818-1mg |
1 mg |
£695.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30048043
|
Novus Biologicals™
NBP3-28818-100ug |
100 μg |
£257.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30047707
|
Novus Biologicals™
NBP3-28818-50ug |
50 μg |
£162.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CTLA-4 Monoclonal antibody specifically detects CTLA-4 in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpécification
| CTLA-4 | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Adaptive Immunity, Autoimmune Diseases, Cancer, Immune System Diseases, Immunology, Neuroscience, Signal Transduction | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CD, CD152 antigen, CD152IDDM12, CD28, celiac disease 3, CELIAC3, CTLA-4, cytotoxic T-lymphocyte antigen 4, cytotoxic T-lymphocyte protein 4, Cytotoxic T-lymphocyte-associated antigen 4, cytotoxic T-lymphocyte-associated protein 4, cytotoxic T-lymphocyte-associated serine esterase-4, GRD4, GSE, ICOS, ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4 | |
| CTLA-4/ CD152 | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| Antitope patent anti-CTLA4 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P16410 | |
| 1493 | |
| IgG1 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit