missing translation for 'onlineSavingsMsg'
Learn More

CTLA-4 Antibody (9H10), Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18318922 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18318922 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18318922 Supplier Novus Biologicals Supplier No. NBP311966AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CTLA-4 Monoclonal antibody specifically detects CTLA-4 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTLA-4
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 9H10
Conjugate Alexa Fluor 647
Dilution Western Blot, Flow Cytometry, ELISA
Formulation 50mM Sodium Borate
Gene Alias CD, CD152 antigen, CD152IDDM12, CD28, celiac disease 3, CELIAC3, CTLA-4, cytotoxic T-lymphocyte antigen 4, cytotoxic T-lymphocyte protein 4, Cytotoxic T-lymphocyte-associated antigen 4, cytotoxic T-lymphocyte-associated protein 4, cytotoxic T-lymphocyte-associated serine esterase-4, GRD4, GSE, ICOS, ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4
Host Species Rabbit
Immunogen This antibody was raised by immunising Syrian hamsters with Staphylococcus A bacteria coated in CTLA-4, isolating B cells from the immunised hamsters and fusing these with the P3X3.Ag8.653 myeloma line to produce stable hybridomas.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Autoimmune Diseases, Cancer, Immune System Diseases, Immunology, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1493
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.