missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTLA-4 Antibody (1B8), Janelia Fluor™ 549, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Brand: Novus Biologicals NBP1-41000JF549
This item is not returnable.
View return policy
Description
CTLA-4 Monoclonal specifically detects CTLA-4 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| CTLA-4 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CTLA4 | |
| Extracellular portion of mouse CD152 fused to a mouse IgG2a. | |
| 0.1 mL | |
| Primary | |
| This antibody recognises mouse CD152, commonly known as CTLA-4 (cytotoxic T-lymphocyte protein 4), a structural homolog for CD28. CD152 binds the same ligands as CD28, namely CD80 (B7.1) and CD86 (B7.2), but with much higher affinity. However, while binding of CD28 enhances T cell function, binding of CD152 inhibits it. CD152 is not detected on naive T-cells and is induced upon T-cell activation. | |
| Store at 4°C in the dark. | |
| IgG |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| 1B8 | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| CD, CD152 antigen, CD152IDDM12, CD28, celiac disease 3, CELIAC3, CTLA-4, cytotoxic T-lymphocyte antigen 4, cytotoxic T-lymphocyte protein 4, Cytotoxic T-lymphocyte-associated antigen 4, cytotoxic T-lymphocyte-associated protein 4, cytotoxic T-lymphocyte-associated serine esterase-4, GRD4, GSE, ICOS, ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4 | |
| Hamster | |
| Protein G purified | |
| Adaptive Immunity, Autoimmune Diseases, Cancer, Immune System Diseases, Immunology, Neuroscience, Signal Transduction | |
| 1493 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction