missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ctip1 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-261PE
This item is not returnable.
View return policy
Description
Ctip1 Polyclonal specifically detects Ctip1 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| Ctip1 | |
| Polyclonal | |
| Flow Cytometry | |
| B-cell CLL/lymphoma 11A, B-cell CLL/lymphoma 11A (zinc finger protein), B-cell CLL/lymphoma 11A (zinc finger protein) isoform 2, B-cell lymphoma/leukemia 11A, BCL-11A, BCL11A B-cell CLL/lymphoma 11A (zinc finger protein) isoform 1, BCL11A-S, BCL11A-XL, C2H2-type zinc finger protein, COUP-TF-interacting protein 1, CTIP1ecotropic viral integration site 9, ecotropic viral integration site 9 homolog, Ecotropic viral integration site 9 protein homolog, EVI-9, EVI9FLJ34997, HBFQTL5, KIAA1809BCL11A-L, Zinc finger protein 856, ZNF856FLJ10173 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| NB600-261 is specific for human Bcl-11a protein. | |
| Store at 4C in the dark. |
| Flow Cytometry | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| BCL11A | |
| A synthetic peptide which maps to a region between residues 775 and the C-terminus (residue 835) of human B-cell CLL/Lymphoma 11A using the numbering given in Swiss-Prot entry Q9H165 (GeneID 53335). | |
| 100 μL | |
| Apoptosis, Cancer, Inhibitors Activators and Regulators, Transcription Factors and Regulators, Tumor Suppressors | |
| 53335 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction