missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CtIP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584133
This item is not returnable.
View return policy
Description
Immunogen sequence: FSAIQRQEKS QGSETSKNKF RQVTLYEALK TIPKGFSSSR KASDGNCTLP KDSPGEPCSQ ECIILQPLNK CSPDNKPSLQ IKEENAVFKI PLRPRESLET E Highest antigen sequence indentity to the following orthologs: Rat - 59%, Mouse - 60%.
CtIP, a 125 kD protein, was originally found interacting with a transcription repressor, CtBP, through the PLDLS motif (CtBP-Interacting Protein) thus suggested a role in transcription. Studies have shown that CtIP also interacts with BRCA1 protein through the c-terminus BRCT domains also suggested that CtIP is a potential tumor suppressor. This CtIP-BRCA1 interaction can be disrupted by DNA damaging agents including UV or gamma-irradiation. Li et al (Nature 406, 210 - 215 (2000)) have shown that ATM phosphorylates CtIP at serine residues 664 and 745, and mutation of these sites abrogates the dissociation of BRCA1 from CtIP, resulting in persistent repression of BRCA1-dependent induction of GADD45 upon ionizing radiation.
Specifications
| CtIP | |
| Polyclonal | |
| Unconjugated | |
| Rbbp8 | |
| 9930104E21Rik; COM1; CtBP-interacting protein; CtIP; DNA endonuclease RBBP8; JWDS; RB binding protein 8, endonuclease; RBBP8; RBBP-8; retinoblastoma binding protein 8; retinoblastoma-binding protein 8; Retinoblastoma-interacting protein and myosin-like; RGD1308872; RIM; SAE2; SCKL2; Sporulation in the absence of SPO11 protein 2 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5932 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.40 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q99708 | |
| Rbbp8 | |
| Recombinant protein corresponding to Human RBBP8. Recombinant protein control fragment (Product #RP-106429). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction