missing translation for 'onlineSavingsMsg'
Learn More

CtIP Antibody, Novus Biologicals™

Product Code. 18410682 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.10mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18410682 25 μL 25µL
18478131 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18410682 Supplier Novus Biologicals Supplier No. NBP18538425ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CtIP Polyclonal antibody specifically detects CtIP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CtIP
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2) and 40% Glycerol
Gene Alias CtBP-interacting protein, CtIP, DNA endonuclease RBBP8, EC 3.1, RBBP-8, retinoblastoma binding protein 8, Retinoblastoma-binding protein 8, Retinoblastoma-interacting protein and myosin-like, RIMSAE2, Sporulation in the absence of SPO11 protein 2 homolog
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: ELECEEDVIPDSPITAFSFSGVNRLRRKENPHVRYIEQTHTKLEHSVCANEMRKVSKSSTHPQHNPNENEILVADTYDQSQSPMAKAHGTSSYTPDKS
Purification Method Immunogen affinity purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 5932
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.