missing translation for 'onlineSavingsMsg'
Learn More

CtIP Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18788240 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18788240 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18788240 Supplier Novus Biologicals Supplier No. NB10079809AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CtIP Polyclonal antibody specifically detects CtIP in Human, Mouse samples. It is validated for Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen CtIP
Applications Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias CtBP-interacting protein, CtIP, DNA endonuclease RBBP8, EC 3.1, RBBP-8, retinoblastoma binding protein 8, Retinoblastoma-binding protein 8, Retinoblastoma-interacting protein and myosin-like, RIMSAE2, Sporulation in the absence of SPO11 protein 2 homolog
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residues 525 and 575 of human CtBP Interacting Protein (Retinoblastoma Binding Protein 8) using the numbering given in entry NP_002885.1 (GeneID 5932).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 5932
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.