missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTHRC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16534-1-AP-20UL
This item is not returnable.
View return policy
Description
The Collagen triple helix repeat-containing protein (CTHRC1) is a glycosylated, secreted protein that is transiently expressed in the arterial wall in response to injury where it is thought to contribute to vascular remodeling by inhibiting collagen expression and deposition and promoting cell migration. CTHRC1 is highly expressed multiple human cancers, including pancreatic cancer where it plays a significant role in the progression and metastasis of the disease by regulating migration and adhesion of tumors cells. CTHRC1 overexpression can also be used as an independent prognostic marker in gastric cancer. Mutations at this locus have been associated with Barrett esophagus and esophageal adenocarcinoma.Specifications
| CTHRC1 | |
| Polyclonal | |
| Unconjugated | |
| CTHRC1 | |
| CTHRC1, Protein NMTC1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 115908, 282836, 68588 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8CG08, Q96CG8, Q9D1D6 | |
| Cthrc1 | |
| CTHRC1 Fusion Protein Ag9812 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction