missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CTHRC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566434
This item is not returnable.
View return policy
Description
Immunogen sequence: QCSWSSLNYGI DLGKIAECTF TKMRSNSALR VLFSGSLRLK CRNACCQRWY FTFNGAECSG PLPI Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
The Collagen triple helix repeat-containing protein (CTHRC1) is a glycosylated, secreted protein that is transiently expressed in the arterial wall in response to injury where it is thought to contribute to vascular remodeling by inhibiting collagen expression and deposition and promoting cell migration. CTHRC1 is highly expressed multiple human cancers, including pancreatic cancer where it plays a significant role in the progression and metastasis of the disease by regulating migration and adhesion of tumors cells. CTHRC1 overexpression can also be used as an independent prognostic marker in gastric cancer. Mutations at this locus have been associated with Barrett esophagus and esophageal adenocarcinoma.
Specifications
| CTHRC1 | |
| Polyclonal | |
| Unconjugated | |
| Cthrc1 | |
| 1110014B07Rik; collagen triple helix repeat containing 1; collagen triple helix repeat-containing protein 1; CTHRC1; Protein NMTC1; UNQ762/PRO1550 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 115908 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96CG8 | |
| Cthrc1 | |
| Recombinant Human CTHRC1. Recombinant protein control fragment (Product #RP-107108). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction