missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CTHRC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA538054
This item is not returnable.
View return policy
Description
A recommended positive control is rat small intestine tissue lysate. At least two isoforms of CTHRC1 are known to exist; this antibody will detect only the larger isoform.
The Collagen triple helix repeat-containing protein (CTHRC1) is a glycosylated, secreted protein that is transiently expressed in the arterial wall in response to injury where it is thought to contribute to vascular remodeling by inhibiting collagen expression and deposition and promoting cell migration. CTHRC1 is highly expressed multiple human cancers, including pancreatic cancer where it plays a significant role in the progression and metastasis of the disease by regulating migration and adhesion of tumors cells. CTHRC1 overexpression can also be used as an independent prognostic marker in gastric cancer. Mutations at this locus have been associated with Barrett esophagus and esophageal adenocarcinoma.
Specifications
| CTHRC1 | |
| Polyclonal | |
| Unconjugated | |
| Cthrc1 | |
| 1110014B07Rik; collagen triple helix repeat containing 1; collagen triple helix repeat-containing protein 1; CTHRC1; Protein NMTC1; UNQ762/PRO1550 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 115908, 282836, 68588 | |
| Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q8CG08, Q96CG8, Q9D1D6 | |
| Cthrc1 | |
| 19 amino acid peptide near the amino terminus of human CTHRC1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction