missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CTHRC1 Polyclonal antibody specifically detects CTHRC1 in Human samples. It is validated for ELISA
Specifications
Specifications
| Formulation | 50mM Sodium Borate |
| Gene Alias | collagen triple helix repeat containing 1, collagen triple helix repeat-containing protein 1, Protein NMTC1 |
| Research Discipline | Cancer |
| Target Species | Human |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?