missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTHRC1 Antibody (166), DyLight 755, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89947IR
This item is not returnable.
View return policy
Description
CTHRC1 Monoclonal specifically detects CTHRC1 in Human samples. It is validated for Western Blot.
Specifications
| CTHRC1 | |
| Monoclonal | |
| DyLight 755 | |
| collagen triple helix repeat containing 1, collagen triple helix repeat-containing protein 1, Protein NMTC1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human CTHRC1 (NP_612464.1; Met1-Lys243). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 166 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 115908 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction