missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CTH Recombinant Rabbit Monoclonal Antibody (23GB3805), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552635
This item is not returnable.
View return policy
Description
This gene encodes a cytoplasmic enzyme in the trans-sulfuration pathway that converts cystathione derived from methionine into cysteine. Glutathione synthesis in the liver is dependent upon the availability of cysteine. Mutations in this gene cause cystathioninuria. Alternative splicing of this gene results in two transcript variants encoding different isoforms.
Specifications
| CTH | |
| Recombinant Monoclonal | |
| Unconjugated | |
| CTH | |
| 0610010I13Rik; AI314617; BC019483; CSE; Cth; cth protein; CTL target antigen; Cys3; cystathionase; cystathionase (cystathionine gamma-lyase); cystathionase isoform 1; cystathionine gamma-lyase; Cystathionine gamma-lyase-like protein; cysteine desulfhydrase; cysteine-protein sulfhydrase; EGK_00877; fb58d08; gamma-cystathionase; H; homoserine deaminase; homoserine dehydratase; LOC102117863; MGC9471; PRB-RA; probasin-related antigen; QccE-22305; synthase; Unknown (protein for MGC:85785); wu:fb48e03; wu:fb58d08; zgc:66001; zgc:85785 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1491 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB3805 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P32929 | |
| CTH | |
| A synthetic peptide of human Cystathionase (1-55AA). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction