missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTGF Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | CTGF |
|---|---|
| Concentration | 700 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17609616
|
Proteintech
25474-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17699606
|
Proteintech
25474-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CTGF Polyclonal antibody apecifically detetects CTGF antigen in Human, Rat Samples. It is validated for Western Blot, ELISA.Specifications
| CTGF | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| 1490 | |
| Peptide | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 700 μg/mL | |
| Polyclonal | |
| Liquid | |
| Human, Rat | |
| NM_001901 | |
| CTGF | |
| IgG | |
| Antigen affinity purification |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title