missing translation for 'onlineSavingsMsg'
Learn More

CTGF/CCN2 Antibody (28711), Novus Biologicals™

Product Code. 18386245 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18386245 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18386245 Supplier Novus Biologicals Supplier No. NBP313686

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CTGF/CCN2 Monoclonal antibody specifically detects CTGF/CCN2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTGF/CCN2
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 28711
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry, Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS, 20% Glycerol
Gene Alias CCN2IGFBP-8, connective tissue growth factor, HCS24, Hypertrophic chondrocyte-specific protein 24, IBP-8, IGF-binding protein 8, IGFBP8CCN family member 2, Insulin-like growth factor-binding protein 8, MGC102839, NOV2
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the center region of human CTGF/CCN2. The exact sequence is proprietary.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Biology, Cytokine Research, Growth and Development, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 1490
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.