missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SCP3 Monoclonal Antibody (6F9C5)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA524798

Product Code. 15924444

  • £453.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SCP3 Monoclonal Antibody for Western Blot, IHC (P), ICC/IF, Flow, ELISA

This gene encodes an essential structural component of the synaptonemal complex. This complex is involved in synapsis, recombination and segregation of meiotic chromosomes. Mutations in this gene are associated with azoospermia in males and susceptibility to pregnancy loss in females. Alternate splicing results in multiple transcript variants that encode the same protein.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SCP3
Monoclonal
1 mg/mL
PBS with 0.5% proprietary stabilizer and 0.05% sodium azide
Q8IZU3
Sycp3
Partial recombinant human SCP3 (between residues 1-200) expressed in E. coli.
100 μL
Primary
Human
Antibody
IgG1
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
6F9C5
Unconjugated
Sycp3
Cor1; LOC615896 protein; RNASCP3; RPRGL4; Scp3; SCP-3; SPGF4; Sycp3; Synaptonemal complex protein 3; synaptonemal complex protein 3-like
Mouse
Ammonium sulfate precipitation
RUO
50511
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.