missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTDSP2 Monoclonal Antibody (4F4)

CTDSP2 Monoclonal Antibody (4F4)
Brand: Invitrogen MA520929
This item is not returnable.
View return policy
Description
Specifications
| CTDSP2 | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| O14595 | |
| CTDSP2 | |
| CTDSP2 (AAH65920, 1 a.a. ~ 271 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 4F4 | |
| Unconjugated | |
| CTDSP2 | |
| AI586070; Carboxy-terminal domain RNA polymerase II polypeptide A small phosphatase 2; conserved gene amplified in osteosarcoma; CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase 2; CTD small phosphatase 2; CTDSP2; D10Ertd73e; NIF2; NLI-interacting factor 2; nuclear LIM interactor-interacting factor 2; nuclear LIM interactor-interacting factor 2 (NLI-interacting factor 2) (Protein OS-4); nuclear LIM interactor-interacting factor 2-like; OS4; OS-4; protein OS-4; PSR2; SCP2; small CTD phosphatase 2; Small C-terminal domain phosphatase 2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10106 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction