missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTDSP2 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97362JF549
This item is not returnable.
View return policy
Description
CTDSP2 Polyclonal specifically detects CTDSP2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| CTDSP2 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Janelia Fluor 549 | |
| carboxy-terminal domain RNA polymerase II polypeptide A small phosphatase 2, conserved gene amplified in osteosarcoma, CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) smallphosphatase 2, NIF2, Nuclear LIM interactor-interacting factor 2, OS4NLI-interacting factor 2, Protein OS-4, PSR2, SCP2EC 3.1.3.16, Small CTD phosphatase 2, Small C-terminal domain phosphatase 2 | |
| Produced in rabbits immunized with E. coli-derived Human CTDSP2 fragment. | |
| 0.1 mL | |
| Protein Phosphatase | |
| 10106 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction