missing translation for 'onlineSavingsMsg'
Learn More

CTDSP1 Antibody (OTI1B6), DyLight 594, Novus Biologicals™

Product Code. 18423077 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18423077 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18423077 Supplier Novus Biologicals Supplier No. NBP272110DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CTDSP1 Monoclonal antibody specifically detects CTDSP1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTDSP1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI1B6
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias carboxy-terminal domain RNA polymerase II polypeptide A small phosphatase 1, CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) smallphosphatase 1, NIF3, NLI-IF, NLIIFEC 3.1.3.16, NLI-interacting factor 3, SCP1Nuclear LIM interactor-interacting factor 3, Small CTD phosphatase 1, Small C-terminal domain phosphatase 1
Host Species Mouse
Immunogen Full length recombinant protein of human CTDSP1 (NP_067021) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 58190
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.