missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTDSP1 Antibody (OTI1B6), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72110CL1
This item is not returnable.
View return policy
Description
CTDSP1 Monoclonal antibody specifically detects CTDSP1 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot
Specifications
| CTDSP1 | |
| Monoclonal | |
| CoraFluor 1 | |
| carboxy-terminal domain RNA polymerase II polypeptide A small phosphatase 1, CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) smallphosphatase 1, NIF3, NLI-IF, NLIIFEC 3.1.3.16, NLI-interacting factor 3, SCP1Nuclear LIM interactor-interacting factor 3, Small CTD phosphatase 1, Small C-terminal domain phosphatase 1 | |
| Full length human recombinant protein of human CTDSP1 (NP_067021) produced in E.coli. | |
| 0.1 mL | |
| Signal Transduction | |
| 58190 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Western Blot | |
| OTI1B6 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction