missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CTCF Recombinant Rabbit Monoclonal Antibody (23GB3520), Invitrogen™
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£179.00 - £485.00

Specifications

Antigen CTCF
Clone 23GB3520
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30089363
GREENER_CHOICE
View Documents
Invitrogen™
MA553394
100 μL
£485.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30089402
View Documents
Invitrogen™
MA553395
20 μL
£179.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CTCF Recombinant Monoclonal antibody specifically detects CTCF in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot
Specifications

Specifications

CTCF
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
Human, Mouse, Rat
P49711, Q61164, Q9R1D1
10664, 13018, 83726
IgG
Affinity chromatography
Antibody
23GB3520
Recombinant Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
11 zinc finger transcriptional repressor; 11-zinc finger protein; AW108038; CCCTC-binding factor; CCCTC-binding factor (zinc finger protein); Ctcf; ctcf protein; CTCFL paralog; im:7152666; MRD21; OTTHUMP00000197610; Transcriptional repressor CTCF; Unknown (protein for IMAGE:8125283)
A synthetic peptide of human CTCF (680-727AA).
Primary
-20°C
CTCF
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.