missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTCF, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova PAB31287.50ug
This item is not returnable.
View return policy
Specifications
| CTCF | |
| Polyclonal | |
| ELISA (1:10000) Western Blot (1:1000) ChIP (1 ug/CHIP) Immunofluorescence (1:200) The optimal working dilution should be determined by the end user. | |
| CCCTC-binding factor (zinc finger protein) | |
| CTCF | |
| A synthetic peptide (conjugated with KLH) corresponding to human CTCF. | |
| RUO | |
| 10664 | |
| Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| ChIP Assay, ChIP sequencing (ChIP-seq), ELISA, Immunofluorescence, Western Blot | |
| Unconjugated | |
| In PBS (0.05% sodium azide, 0.05% proclin 300). | |
| - | |
| Rabbit | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction