Learn More
Invitrogen™ CTCF Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588115
Description
Immunogen sequence: MEGDAVEAIV EESETFIKGK ERKTYQRRRE GGQEEDACHL PQNQTDGGEV VQDVNSSVQM VMMEQLDPTL LQMKTEVMEG TVAPEAEAAV DDTQIITLQV VNMEEQPINI GELQLVQVPV PVTVPVATTS VEELQGAYEN EVSKEGLAES EPMICHTLPL PEGFQVVKVG ANGEVETLEQ GELPPQEDPS WQKDPDYQPP AKKTKKTKKS KLRYTEEGKD VDVSVYDFEE EQQEGLLSEV NAEKVVGNMK PPKPTKIKKK; Positive Samples: 293T, Jurkat; Cellular Location: Chromosome, Nucleus, centromere, nucleoplasm.
Transcriptional factor CTCF is a ubiquitously expressed 11-zinc finger domain containing, DNA-binding nuclear phosphoprotein that is involved in multiple aspects of normal gene regulation including transcriptional repression and activation, gene silencing, chromatin insulation and regulation of imprinted sites. CTCF uses different combinations of ZF domains and interacts with the CCCTC motif on the DNA. It can bind to HAT- and HDAC-containing complexes thereby influencing transcriptional activation or repression respectively. It is a multifunctional protein involved in the transactivation of APPB gene, silencing of c-myc gene, and insulation of human beta-globin and DMI myotonic dystrophy locus and imprinting control of IGF2 and H19 regions. CTCF has also been shown to regulate the transcription of Pax6 and IRAK2 promoter thereby opening a therapeutic avenue. Mutations in this gene have been associated with invasive breast cancers, prostate cancers and Wilms' tumors.
Specifications
| CTCF | |
| Polyclonal | |
| Unconjugated | |
| CTCF | |
| 11 zinc finger transcriptional repressor; 11-zinc finger protein; AW108038; CCCTC-binding factor; CCCTC-binding factor (zinc finger protein); Ctcf; ctcf protein; CTCFL paralog; im:7152666; MRD21; OTTHUMP00000197610; Transcriptional repressor CTCF; Unknown (protein for IMAGE:8125283) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10664, 13018, 83726 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ChIP Assay, ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P49711, Q61164, Q9R1D1 | |
| CTCF | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human CTCF (NP_0065561). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.