missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTCF Antibody (CL0307) [Janelia Fluor™ 646], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52911JF646
This item is not returnable.
View return policy
Description
CTCF Monoclonal antibody specifically detects CTCF in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| CTCF | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 11 zinc finger transcriptional repressor, 11-zinc finger protein, CCCTC-binding factor, CCCTC-binding factor (zinc finger protein), CTCFL paralog, transcriptional repressor CTCF | |
| This antibody was developed using a recombinant protein derived from P49711, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Immune System Diseases, Immunology, Transcription Factors and Regulators | |
| 10664 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL0307 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction