missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTBS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12599-1-AP-20UL
This item is not returnable.
View return policy
Description
Involved in the degradation of asparagine-linked glycoproteins. Hydrolyze of N-acetyl-beta-D-glucosamine (1-4)N-acetylglucosamine chitobiose core from the reducing end of the bond, it requires prior cleavage by glycosylasparaginase.Specifications
| CTBS | |
| Polyclonal | |
| Unconjugated | |
| CTBS | |
| chitobiase, di N acetyl, CTB, CTBS, Di N acetylchitobiase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1486, 74245, 81652 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q01459, Q01460, Q8R242 | |
| Ctbs | |
| CTBS Fusion Protein Ag3228 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction