missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CtBP1 Recombinant Rabbit Monoclonal Antibody (23GB5115), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552794
This item is not returnable.
View return policy
Description
This gene encodes a protein that binds to the C-terminus of adenovirus E1A proteins. This phosphoprotein is a transcriptional repressor and may play a role during cellular proliferation. This protein and the product of a second closely related gene, CTBP2, can dimerize. Both proteins can also interact with a polycomb group protein complex which participates in regulation of gene expression during development. Alternative splicing of transcripts from this gene results in multiple transcript variants.
Specifications
| CtBP1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| CTBP1 | |
| 50 kDa BFA-dependent ADP-ribosylation substrate; 50-kDa BFA-induced ADP-ribosylated substrate; 50-kDaBFA-inducedADP-ribosylatedsubstrate; BARS; BARS-50; brefeldin A-ADP-riboslyated substrate; brefeldin A-ribosylated substrate; CTBP; CtBP1; CtBP1-L; CtBP1-S; CtBP3; CtBP3/BARS; C-terminal binding protein 1; C-terminal-binding protein 1; C-terminal-binding protein 3; C-terminus binding protein 3/brefeldin A (BFA) adenosine diphosphate-ribosylated substrate; D4S115h; D5H4S115; D5H4S115E; MGC104684 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 13016, 1487, 29382 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5115 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O88712, Q13363, Q9Z2F5 | |
| CTBP1 | |
| A synthesized peptide derived from human CtBP1 (350-400AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction