missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CtBP1 Polyclonal Antibody
CtBP1 Polyclonal Antibody
Brand: Invitrogen PA599321
This item is not returnable.
View return policy
Description
CtBP1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
| CtBP1 | |
| Polyclonal | |
| Unconjugated | |
| CTBP1 | |
| 50 kDa BFA-dependent ADP-ribosylation substrate; 50-kDa BFA-induced ADP-ribosylated substrate; 50-kDaBFA-inducedADP-ribosylatedsubstrate; BARS; BARS-50; brefeldin A-ADP-riboslyated substrate; brefeldin A-ribosylated substrate; CTBP; CtBP1; CtBP1-L; CtBP1-S; CtBP3; CtBP3/BARS; C-terminal binding protein 1; C-terminal-binding protein 1; C-terminal-binding protein 3; C-terminus binding protein 3/brefeldin A (BFA) adenosine diphosphate-ribosylated substrate; D4S115h; D5H4S115; D5H4S115E; MGC104684 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 13016, 1487, 29382 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O88712, Q13363, Q9Z2F5 | |
| CTBP1 | |
| A synthesized peptide derived from human CTBP1(Accession Q13363), corresponding to amino acid residues T80-D130. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction