missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CtBP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA110028
This item is not returnable.
View return policy
Description
This antibody reacts with CtBP1, a 49 kDa transcriptional corepressor (intracellular antigen), that plays a critical role in regulating gene expression. Western Blot: Reducing conditions; Immunocytochemistry: acetone/methanol fixation.
This gene encodes a protein that binds to the C-terminus of adenovirus E1A proteins. This phosphoprotein is a transcriptional repressor and may play a role during cellular proliferation. This protein and the product of a second closely related gene, CTBP2, can dimerize. Both proteins can also interact with a polycomb group protein complex which participates in regulation of gene expression during development. Alternative splicing of transcripts from this gene results in multiple transcript variants.
Specifications
| CtBP1 | |
| Polyclonal | |
| Unconjugated | |
| CTBP1 | |
| 50 kDa BFA-dependent ADP-ribosylation substrate; 50-kDa BFA-induced ADP-ribosylated substrate; 50-kDaBFA-inducedADP-ribosylatedsubstrate; BARS; BARS-50; brefeldin A-ADP-riboslyated substrate; brefeldin A-ribosylated substrate; CTBP; CtBP1; CtBP1-L; CtBP1-S; CtBP3; CtBP3/BARS; C-terminal binding protein 1; C-terminal-binding protein 1; C-terminal-binding protein 3; C-terminus binding protein 3/brefeldin A (BFA) adenosine diphosphate-ribosylated substrate; D4S115h; D5H4S115; D5H4S115E; MGC104684 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 13016, 1487 | |
| 4°C, do not freeze | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 15mM sodium azide; pH 7.4 | |
| O88712, Q13363 | |
| CTBP1 | |
| Bacterially expressed full-length human CtBP1. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction