missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTBP1-DT Antibody (OTI4F2) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72334
This item is not returnable.
View return policy
Description
CTBP1-DT Monoclonal specifically detects CTBP1-DT in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| C4orf42 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:2000, Flow Cytometry 1:100, CyTOF-ready | |
| CTBP1 antisense RNA 1 | |
| Mouse | |
| 19.5 kDa | |
| 100 μg | |
| 92070 | |
| Human | |
| Lyophilized |
| Western Blot, Flow Cytometry, CyTOF | |
| OTI4F2 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| CTBP1-DT | |
| Human recombinant protein fragment corresponding to amino acids 1-186 of human C4orf42(NP_443093) produced in E.coli. | |
| Affinity Purified | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction