missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTBP1-DT Antibody (OTI4F2), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72334AF488
This item is not returnable.
View return policy
Description
CTBP1-DT Monoclonal specifically detects CTBP1-DT in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| CTBP1-DT | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CTBP1-DT | |
| Recombinant protein fragment corresponding to amino acids 1-186 of human C4orf42 (NP_443093) produced in E.coli. | |
| 0.1 mL | |
| 92070.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry | |
| OTI4F2 | |
| Western Blot, Flow Cytometry | |
| CTBP1 antisense RNA 1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction