Learn More
CtBP1 Antibody (4D6), mFluor Violet 450 SE, Novus Biologicals™
Product Code. 18424528 Shop All Bio Techne Products
Product Code. 18424528 Supplier Novus Biologicals Supplier No. NBP174040MFV450
Mouse Monoclonal Antibody
CtBP1 Monoclonal antibody specifically detects CtBP1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence