Learn More
Invitrogen™ CTACK (CCL27A) Mouse ELISA Kit
Sandwich ELISA Kit
£504.00 - £2310.00
Specifications
Accession Number | Q9Z1X0 |
---|---|
Assay Sensitivity | 15 pg/mL |
Assay | Solid-phase sandwich ELISA |
Conjugate | Biotin |
Product Type | ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15465233
|
Invitrogen™
EMCCL27A |
96 Tests |
£504.00
1 set |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
17115699
|
Invitrogen™
EMCCL27AX5 |
5 x 96 Tests |
£2310.00
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
Description
The Mouse CTACK (CCL27A) ELISA quantitates Ms CTACK in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms CTACK. Principle of the method The Mouse CTACK solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
This protein is one of several CC cytokine genes clustered on the p-arm of chromosome 9. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cyste.Specifications
Q9Z1X0 | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
Mouse | |
Colorimetric Microplate Reader | |
ALP,AW558992,CTACK,CTAK,Ccl27,ESkine,ILC,PESKY,Scya27,Scya27a | |
<10% | |
RUO | |
2°C to 8°C | |
Mouse | |
4 hr. 45 min. |
15 pg/mL | |
Biotin | |
Chemokine & Receptors | |
This ELISA kit shows no cross-reactivity with any of the cytokines tested: Mouse CD30, L CD30, T CD40, CRG-2, CXCL16, Eotaxin , Eotaxin-2, Fas Ligand, Fractalkine, GCSF, GM-CFS, IFN- gamma, IGFBP-2, IGFBP-3, IGFBP-5, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-3 Rb, IL-4, IL-5, IL-6, IL-9, IL-10, IL-12 p40/p70, IL-12 p70, IL-13, IL-17, KC, Leptin R, LEPTIN(OB), LIX, L-Selectin, Lymphotactin, MCP-1, MCP-5, M-CSF, MIG, MIP-1 alpha, MIP-1 gamma, MIP-2, MIP-3 beta, MIP-3 alpha, PF-4, P-Selectin, RANTES, SCF, SDF-1 alpha, TARC, TCA-3, TECK, TIMP-1, TNF-alpha, TNF RI, TNF RII, TPO, VCAM-1, VEGF. | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
20301 | |
<12% | |
HRP | |
Immunology | |
RP23-167I12.8, ALP, AW558992, CTACK, CTAK, Ccl27, ESkine, ILC, PESKY, Scya27, Scya27a | |
1 hr. 20 min. |