missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CT45A3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92094-0.1ml
This item is not returnable.
View return policy
Description
CT45A3 Polyclonal antibody specifically detects CT45A3 in Mouse, Rat samples. It is validated for Western Blot
Specifications
| CT45A3 | |
| Polyclonal | |
| Western Blot 1:1000-1:2000 | |
| Cancer/testis antigen 45-3, Cancer/testis antigen 45A3, cancer/testis antigen CT45-3, cancer/testis antigen family 45 member A3, cancer/testis antigen family 45, member A3, CT45-3CT45.3RP13-36C9.1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-189 of human CT45A3 (NP_001017435.1). MTDKTEKVAVDPETVFKRPRECDSPSYQKRQRMALLARKQGAGDSLIAGSAMSKEKKLMTGHAIPPSQLDSQIDDFTGFSKDRMMQKPGSNAPVGGNVTSSFSGDDLECRETASSPKSQREINADIKRKLVKELRCVGQKYEKIFEMLEGVQGPTAVRKRFFESIIKEAARCMRRDFVKHLKKKLKRMI | |
| 0.1 mL | |
| Cancer, Immunology | |
| 441519 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction