missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CT45A Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560807
This item is not returnable.
View return policy
Description
Immunogen sequence: TEKVAVDPET VFKRPRECDS PSYQKRQRMA LLARKQGAGD SLIAGSAMSK Highest antigen sequence identity to the following orthologs: Mouse - 30%, Rat - 30%.
CT45A3 (Cancer/Testis Antigen Family 45 Member A3) is a Protein Coding gene. An important paralog of this gene is CT45A1.
Specifications
| CT45A | |
| Polyclonal | |
| Unconjugated | |
| CT45A5 | |
| cancer/testis antigen 45-1; Cancer/testis antigen 45-3; cancer/testis antigen 45-4; cancer/testis antigen 45-4/6; cancer/testis antigen 45-5; Cancer/testis antigen 45-6; cancer/testis antigen 45A1; Cancer/testis antigen 45A10; cancer/testis antigen 45A3; Cancer/testis antigen 45A4; cancer/testis antigen 45A4/45A6; cancer/testis antigen 45A5; Cancer/testis antigen 45A6; Cancer/testis antigen 45A7; cancer/testis antigen CT45-1; cancer/testis antigen CT45-3; cancer/testis antigen CT45-5; cancer/testis antigen CT45-6; cancer/testis antigen family 45 member A1; Cancer/testis antigen family 45 member A10; cancer/testis antigen family 45 member A3; Cancer/testis antigen family 45 member A4; cancer/testis antigen family 45 member A4/A6; cancer/testis antigen family 45 member A5; Cancer/testis antigen family 45 member A6; Cancer/testis antigen family 45 member A7; cancer/testis antigen family 45 member A-like; cancer/testis antigen family 45, member A1; cancer/testis antigen family 45, member A10; cancer/testis antigen family 45, member A3; cancer/testis antigen family 45, member A4; cancer/testis antigen family 45, member A5; cancer/testis antigen family 45, member A6; CT45; CT45.1; CT45.3; CT45.4; CT45.5; CT45.6; CT45-1; CT45-3; CT45-4; CT455; CT45-5; CT45-6; CT45A1; CT45A10; CT45A3; CT45A4; CT45A5; CT45A6; CT45A7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 102723631, 441519, 441521, 541465, 541466 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P0DMU7, P0DMU8, P0DMU9, Q5HYN5, Q8NHU0 | |
| CT45A1, CT45A10, CT45A3, CT45A5, CT45A6 | |
| Recombinant protein corresponding to Human CT45A1. Recombinant protein control fragment (Product #RP-104370). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction