missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CT45A Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA561395

Product Code. 15749211

  • £476.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: PGSNAPVGGN VTSSFSGDDL ECRETASSPK SQREINADI Highest antigen sequence identity to the following orthologs: Mouse - 37%, Rat - 37%.

CT45A3 (Cancer/Testis Antigen Family 45 Member A3) is a Protein Coding gene. An important paralog of this gene is CT45A1.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CT45A
Polyclonal
Unconjugated
CT45A5
cancer/testis antigen 45-1; Cancer/testis antigen 45-3; cancer/testis antigen 45-4; cancer/testis antigen 45-4/6; cancer/testis antigen 45-5; Cancer/testis antigen 45-6; cancer/testis antigen 45A1; Cancer/testis antigen 45A10; cancer/testis antigen 45A3; Cancer/testis antigen 45A4; cancer/testis antigen 45A4/45A6; cancer/testis antigen 45A5; Cancer/testis antigen 45A6; Cancer/testis antigen 45A7; cancer/testis antigen CT45-1; cancer/testis antigen CT45-3; cancer/testis antigen CT45-5; cancer/testis antigen CT45-6; cancer/testis antigen family 45 member A1; Cancer/testis antigen family 45 member A10; cancer/testis antigen family 45 member A3; Cancer/testis antigen family 45 member A4; cancer/testis antigen family 45 member A4/A6; cancer/testis antigen family 45 member A5; Cancer/testis antigen family 45 member A6; Cancer/testis antigen family 45 member A7; cancer/testis antigen family 45 member A-like; cancer/testis antigen family 45, member A1; cancer/testis antigen family 45, member A10; cancer/testis antigen family 45, member A3; cancer/testis antigen family 45, member A4; cancer/testis antigen family 45, member A5; cancer/testis antigen family 45, member A6; CT45; CT45.1; CT45.3; CT45.4; CT45.5; CT45.6; CT45-1; CT45-3; CT45-4; CT455; CT45-5; CT45-6; CT45A1; CT45A10; CT45A3; CT45A4; CT45A5; CT45A6; CT45A7
Rabbit
Antigen affinity chromatography
RUO
102723631, 441519, 441521, 541465, 541466
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P0DMU7, P0DMU8, P0DMU9, Q5HYN5, Q8NHU0
CT45A1, CT45A10, CT45A3, CT45A5, CT45A6
Recombinant protein corresponding to Human CT45A. Recombinant protein control fragment (Product #RP-104418).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.