missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CT173 Polyclonal Antibody

CT173 Polyclonal Antibody
Brand: Invitrogen PA524315
This item is not returnable.
View return policy
Description
CT173 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
| CT173 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q96LM9 | |
| C20orf173 | |
| KLH conjugated synthetic peptide between 48-76 amino acids from the central region of human CT173. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| C20orf173 | |
| C20orf173; chromosome 20 open reading frame 173; dJ477O4.4; Uncharacterized protein C20orf173 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 140873 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction