missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CT110 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92349-0.02ml
This item is not returnable.
View return policy
Description
CT110 Polyclonal antibody specifically detects CT110 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| CT110 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| cancer/testis antigen 110, CT110, FLJ10252, FLJ21048, G patch domain containing 2, G patch domain-containing protein 2, GPATC2, MGC74998, RP11-361K17.1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human GPATCH2 (NP_001284683.1). MFGAAGRQPIGAPAAGNSWHFSRTMEELVHDLVSALEESSEQARGGFAETGDHSRSISCPLKRQARKRRGRKRRSYNVHHPWETGHCLSEGSDSSLEEPSKDYRENHNNNKKDHSDSDDQMLVAKRRPSSNLNNNVRGKRPLWHESDFAVDNVGNRTLRRRRKVKRMAVDLPQDISNKRTMTQPPEGCRDQDMDSDRAYQYQEFTKNKVKKRKLKIIRQGPKIQDEGVVLESEETNQTNKDKMECEEQKV | |
| 0.02 mL | |
| Cancer, Immunology | |
| 55105 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction