missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ CT Supplement

Product Code. 10787083 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1000 tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10787083 1000 tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10787083 Supplier Applied Biosystems™ Supplier No. 74001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

CT Supplement

CT Supplement is added to standard Sorbitol MacConkey media (SMAC) to form CT-SMAC. CT-SMAC selects for E.coli 0157 after IMS.

This Product Contains: 1mg Cefixime powder and 50mg Potassium Tellurite powder.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator (2–8°C).
Form Powder
Media Type Pathogen Detection Media
Preparation Method Prepare Stock Solution
Target Organism Class E. coli
Final Product Type Liquid Supplement
Quantity 1000 tests
Shipping Condition Room Temperature
Strain Designations E. coli O157

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.