missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CSTF3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588201
This item is not returnable.
View return policy
Description
Immunogen sequence: MSGDGATEQA AEYVPEKVKK AEKKLEENPY DLDAWSILIR EAQNQPIDKA RKTYERLVAQ FPSSGRFWKL YIEAEVTILF YFFLYQYCSI HCSDRKQVRN IAN; Positive Samples: Mouse liver, Rat liver; Cellular Location: Nucleus.
The protein encoded by this gene is one of three (including CSTF1 and CSTF2) cleavage stimulation factors that combine to form the cleavage stimulation factor complex (CSTF). This complex is involved in the polyadenylation and 3' end cleavage of pre-mRNAs. The encoded protein functions as a homodimer and interacts directly with both CSTF1 and CSTF2 in the CSTF complex. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
| CSTF3 | |
| Polyclonal | |
| Unconjugated | |
| CSTF3 | |
| 4732468G05Rik; C79532; CF-1 77 kDa subunit; cleavage stimulation factor 77 kDa subunit; Cleavage stimulation factor subunit 3; cleavage stimulation factor subunit 3, isoform 1; cleavage stimulation factor, 3' pre-RNA, subunit 3; cleavage stimulation factor, 3' pre-RNA, subunit 3, 77kD; cleavage stimulation factor, 3' pre-RNA, subunit 3, 77kDa; CSTF 77 kDa subunit; CSTF3; CSTF-77; MGC117398; MGC43001; MGC75122 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1479, 228410, 362178 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.94 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q12996, Q99LI7 | |
| CSTF3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-103 of human CSTF3 (NP_0010286771). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction