missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CSTF3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA558911
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Immunogen sequence: LAFESNIGDL ASILKVEKRR FTAFKEEYEG KETALLVDRY KFMDLYPCSA SELKALGYKD VSRAKLAAII PDPVVAPSIV PVLKDEVDRK PEYPKPDTQQ MIPF Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
The protein encoded by this gene is one of three (including CSTF1 and CSTF2) cleavage stimulation factors that combine to form the cleavage stimulation factor complex (CSTF). This complex is involved in the polyadenylation and 3' end cleavage of pre-mRNAs. The encoded protein functions as a homodimer and interacts directly with both CSTF1 and CSTF2 in the CSTF complex. Alternative splicing results in multiple transcript variants encoding different isoforms.
Spezifikation
| CSTF3 | |
| Polyclonal | |
| Unconjugated | |
| CSTF3 | |
| 4732468G05Rik; C79532; CF-1 77 kDa subunit; cleavage stimulation factor 77 kDa subunit; Cleavage stimulation factor subunit 3; cleavage stimulation factor subunit 3, isoform 1; cleavage stimulation factor, 3' pre-RNA, subunit 3; cleavage stimulation factor, 3' pre-RNA, subunit 3, 77kD; cleavage stimulation factor, 3' pre-RNA, subunit 3, 77kDa; CSTF 77 kDa subunit; CSTF3; CSTF-77; MGC117398; MGC43001; MGC75122 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1479, 228410 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q12996, Q99LI7 | |
| CSTF3 | |
| Recombinant protein corresponding to Human CSTF3. Recombinant protein control fragment (Product #RP-97498). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur