missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ CSTF2T Recombinant Protein Antigen

Product Code. 18606867 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
This item is not returnable. View return policy

Product Code. 18606867

Brand: Novus Biologicals™ NBP249353PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Antibody Competition
Formulation PBS and 1 M Urea, pH 7.4.
Gene ID (Entrez) 23283
Molecular Weight (g/mol) MolecularWeight-theroretical: 23 kDa
Quantity 0.1 mL
Storage Requirements Store at -20°C. Avoid freeze/thaw cycles.
Gene Alias CF-1 64 kDa subunit tau, CF-1 64 kDa subunit tau variant, cleavage stimulation factor 64 kDa subunit tau, Cleavage stimulation factor 64 kDa subunit tau variant, cleavage stimulation factor subunit 2 tau, cleavage stimulation factor subunit 2 tau variant, cleavage stimulation factor, 3' pre-RNA, subunit 2, 64 kDa, tau variant, CSTF 64 kDa subunit tau variant, CstF-64 T, DKFZp434C1013, KIAA0689CSTF 64 kDa subunit tau, tauCstF-64
Gene Symbol CSTF2T
Inhibitor Properties This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-49353. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/mL
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein CSTF2T
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.