missing translation for 'onlineSavingsMsg'
Learn More

CSTF2T Mouse anti-Human, Clone: PCRP-CSTF2T-1A3, Novus Biologicals™

Product Code. 18399101 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18399101 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18399101 Supplier Novus Biologicals Supplier No. NBP30748620UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CSTF2T Monoclonal specifically detects CSTF2T in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Protein Array.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSTF2T
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Peptide Array
Classification Monoclonal
Clone PCRP-CSTF2T-1A3
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin, Protein Array
Formulation 10 mM PBS with 0.05% BSA
Gene Alias CF-1 64 kDa subunit tau, CF-1 64 kDa subunit tau variant, cleavage stimulation factor 64 kDa subunit tau, Cleavage stimulation factor 64 kDa subunit tau variant, cleavage stimulation factor subunit 2 tau, cleavage stimulation factor subunit 2 tau variant, cleavage stimulation factor, 3' pre-RNA, subunit 2, 64kDa, tau variant, CSTF 64 kDa subunit tau variant, CstF-64T, DKFZp434C1013, KIAA0689CSTF 64 kDa subunit tau, tauCstF-64
Host Species Mouse
Immunogen Recombinant full-length human CSTF2T protein
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23283
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2b κ
Show More Show Less
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.