missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CST 530, Phenol
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
CST 530, Phenol is a functionalized IAP (XIAP and cIAP) ligand for PROTAC™ research and development. Supplied with a phenol functional handle for ready conjugation to a linker/target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.Please contact us for SD files of our available Degrader Building Blocks.PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | tert-Butyl ((S)-1-(((S)-1-cyclohexyl-2-((2S,4S)-4-(3-hydroxyphenoxy)-2-(((R)-1,2,3,4-tetrahydronaphthalen-1-yl)carbamoyl)pyrrolidin-1-yl)-2-oxoethyl)amino)-1-oxopropan-2-yl)(methyl)carbamate |
| CAS | 2241489-43-8 |
| Quantity | 50 mg |
| Target | Degrader Building Blocks |
| Molecular Formula | C38H52N4O7 |
| Purity | 0.95 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction