missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CSRP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583620
This item is not returnable.
View return policy
Description
Immunogen sequence: GAGCLSTDTG EHLGLQFQQS PKPARSVTTS NPSKFTAKFG ESEKCPRCGK SVYA Highest antigen sequence indentity to the following orthologs: Rat - 94%, Mouse - 94%.
This gene encodes a member of the CSRP family of LIM domain proteins, which may be involved in regulatory processes important for development and cellular differentiation. The LIM/double zinc-finger motif found in this protein is found in a group of proteins with critical functions in gene regulation, cell growth, and somatic differentiation. Mutations in this gene are thought to cause heritable forms of hypertrophic cardiomyopathy (HCM) and dilated cardiomyopathy (DCM) in humans. Alternatively spliced transcript variants with different 5' UTR, but encoding the same protein, have been found for this gene.
Specifications
| CSRP3 | |
| Polyclonal | |
| Unconjugated | |
| Csrp3 | |
| cardiac LIM domain protein; cardiac LIM protein; Clp; CMD1M; CMH12; CRP3; Csrp3; cysteine and glycine rich protein 3; cysteine and glycine-rich protein 3; cysteine and glycine-rich protein 3 (cardiac LIM protein); cysteine-rich protein 3; LIM domain only 4; LIM domain protein, cardiac; LMO4; MGC14488; MGC61993; MLP; MMLP; Muscle LIM protein; muscle lim protein isoform | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8048 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P50461 | |
| Csrp3 | |
| Recombinant protein corresponding to Human CSRP3. Recombinant protein control fragment (Product #RP-98423). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction